missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde Solution, Contains 10-15% Methanol, Honeywell™
Shop All Honeywell Research Chemicals ProductsDescription
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Specifications
| Melting Point | <-15°C |
| Color | Colorless |
| Density | 1.09g/mL at 25°C |
| pH | 3.5 to 4.5 (at 20°C) |
| Boiling Point | 93°C to 96°C (1013 hPa) |
| Flash Point | 64°C (147.2°F) |
| Odor | Stinging |
| Packaging | Poly Bottle |
| Linear Formula | HCHO |
| Quantity | 4 L |
| Show More |
Safety and Handling
Product Identifier
- Formaldehyde solution
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity Category 3
- H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H335-May cause respiratory irritation.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H370-Causes damage to organs.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contain: formaldehyde, methanol
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?