missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Extra Pure, Solution 37-41%, SLR, Fisher Chemical™

Product Code. p-7073027 Shop All Fisher Chemical Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
500mL
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

Product Code. 10630813

Brand: Fisher Chemical F/1501/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C | CAS: 50-00-0 | CH2O | 30.03 g/mol

CH2O, CAS Number-50-00-0, 2.5L, 97 deg.C, CHEBI:16842, Colorless, 30.02g/mol, WSFSSNUMVMOOMR-UHFFFAOYSA-N, formaldehyde, -15 deg.C, 3274, 30.026, HDPE plastic bottle, 3 to 4.2, Liquid, 712, C-O, > 1.0, 2mbar at 20 deg.C

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-00-0
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O

Specifications

Packaging HDPE plastic bottle
Vapor Pressure 2mbar at 20°C
Vapor Density > 1.0
Physical Form Liquid
Color Colorless
pH 3 to 4.2
Melting Point -15°C
Boiling Point 97°C
Quantity 500 mL
Formula Weight 30.02g/mol
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Formaldehyde solution, 37%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H370-Causes damage to organs.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde, Methylalcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.