Product Code. 30390288
Click to view available options
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
unitSize
30390288
10 mL
10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code.
30390288
Supplier
Toronto Research Chemicals
Supplier No.
TRCF69135210ML
Please sign in to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
Specifications
Chemical Name or Material
Formaldehyde-d2
CAS
1664-98-8
Molecular Formula
C 2H2 O
Quantity
10 mL
Synonym
Deuterated formaldehyde, Formalin-d2
InChI Formula
InChI=1S/CH2O/c1-2/h1H2/i1D2
SMILES
[2H]C(=O)[2H]
IUPAC Name
dideuteriomethanone
Molecular Weight (g/mol)
32.04
Formula Weight
32.0231
Concentration or Composition (by Analyte or Components)
20 %
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?