missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 36.5 to 38%, Reagent grade, For ACS analysis, MP Biomedicals™

Product Code. 11450612 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450612 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11450612 Supplier MP Biomedicals Supplier No. 0219404780
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 50-00-0
Color Clear colorless
Density 1.04g/cm3 at 20°C
Molecular Formula CH2O
MDL Number MFCD00003274
Quantity 100 mL
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
SMILES C=O
IUPAC Name formaldehyde
Molecular Weight (g/mol) 30.03
PubChem CID 712
ChEBI CHEBI:16842
Formula Weight 30.03
Percent Purity 36.5 to 38%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Formaldehyde, For ACS analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.