missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde 2,4-Dinitrophenylhydrazone-d3, TRC

Product Code. 30393884
Change view
Click to view available options
Quantity:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393884 50 mg
30398852 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30393884 Supplier Toronto Research Chemicals Supplier No. TRCF69140250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 259824-50-5
Molecular Formula C7 2H3 H3 N4 O4
Molecular Weight (g/mol) 213.17
Synonym Formaldehyde, (4,6-dinitrophenyl-2,3,5-d3)hydrazone (9CI), Methanal 2,4-dinitro-3,5,6-trideuteriophenylhydrazone, Formaldehyde-2,4-dinitrophenylhydrazone-d3, Formaldehyde-2,4-dinitrophenylhydrazone-3,5,6-D3, Formaldehyde 2,4-Dinitrophenylhydrazone-3,5,6-d3
IUPAC Name 2,3,5-trideuterio-N-(methylideneamino)-4,6-dinitroaniline
SMILES [2H]c1c([2H])c(c([2H])c(c1NN=C)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C7H6N4O4/c1-8-9-6-3-2-5(10(12)13)4-7(6)11(14)15/h2-4,9H,1H2/i2D,3D,4D
Formula Weight 213.0577
Chemical Name or Material Formaldehyde 2,4-Dinitrophenylhydrazone-d3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.