missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde-13C (20% by weight in water), TRC

Product Code. 30403921
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403921 250 mg
30405068 100 mg
30391718 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403921 Supplier Toronto Research Chemicals Supplier No. TRCF691354250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3228-27-1
Molecular Formula 13C H2 O
Molecular Weight (g/mol) 31.02
Synonym Formaldehyde-13C (6CI,7CI,8CI,9CI), Formaldehyde-13C, Methanal-13C, [13C]Formaldehyde
IUPAC Name (1^{13}C)methanone
SMILES [13CH2]=O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/CH2O/c1-2/h1H2/i1+1
Formula Weight 31.0139
Concentration or Composition (by Analyte or Components) 0.2
Chemical Name or Material Formaldehyde-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.