missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Forkhead box I2 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-20919
This item is not returnable.
View return policy
Description
Forkhead box I2 Polyclonal specifically detects Forkhead box I2 in Mouse, Rat samples. It is validated for Peptide ELISA.
Specifications
| Forkhead box I2 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| FLJ46831, forkhead box I2, forkhead box protein I2, homolog of mouse Foxi2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 399823 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| ELISA | |
| 0.5 mg/mL | |
| Peptide ELISA Detection limit 1:8000 | |
| NP_899016.1 | |
| FOXI2 | |
| Peptide with sequence C-GHLIYSRDGTE corresponding to C-Terminus according to Uniprot Mouse NP_899016.1. | |
| 0.1 mg | |
| Primary | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction