Learn More
Lenz Laborglasintrumente Fork Clamps for Ground Joints
Product Code. 16488743
Change view
Click to view available options
For Use With (Equipment):
NS 24/29 Ground Joints
Unit Size:
Pack of 10
Product Code. For Use With (Equipment) unitSize
16488743 NS 24/29 Ground Joints Pack of 10
1 options
Product Code. 16488743 Supplier Lenz Laborglasintrumente Supplier No. 01103524
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Fork clamps for ground joints
  • For conical ground joints
  • Material: stainless steel
  • Set screw must be ordered separately
Specifications
Quantity 10/Pk.
For Use With (Equipment) NS 24/29 Ground Joints