missing translation for 'onlineSavingsMsg'
Learn More

Forchlorfenuron, TRC

Product Code. 30390287
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30390287

Brand: Toronto Research Chemicals TRCF6910501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Forchlorfenuron
CAS 68157-60-8
Molecular Formula C12 H10 Cl N3 O
Quantity 1 g
Synonym Urea, N-(2-chloro-4-pyridinyl)-N'-phenyl-, N-(2-Chloro-4-pyridinyl)-N'-phenylurea, 1-(2-Chloro-4-pyridyl)-3-phenylurea, 4PU30, CN 11-3138, CPPU, Forchlorfenuron, Fulmet, KT 30, KT 30 (plant growth regulator), N-(2-Chloro-4-pyridyl)-N'-phenylurea, N-Phenyl-N'-(2-chloro-4-pyridyl)urea, SKW 20010, Sitofex, V 3183
InChI Formula InChI=1S/C12H10ClN3O/c13-11-8-10(6-7-14-11)16-12(17)15-9-4-2-1-3-5-9/h1-8H,(H2,14,15,16,17)
SMILES Clc1cc(NC(=O)Nc2ccccc2)ccn1
IUPAC Name 1-(2-chloropyridin-4-yl)-3-phenylurea
Molecular Weight (g/mol) 247.68
Formula Weight 247.0512
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.