missing translation for 'onlineSavingsMsg'
Learn More

Forchlorfenuron-d5, TRC

Product Code. 30396335
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396335 2.5 mg 2.5mg
30398813 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30396335 Supplier Toronto Research Chemicals Supplier No. TRCF6910522.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1398065-87-6
Molecular Formula C12 2H5 H5 Cl N3 O
Molecular Weight (g/mol) 252.71
Synonym Forchlorfenuron D5 (phenyl D5), N-(2-Chloro-4-pyridinyl)-N'-(phenyl-d5)urea, 1-(2-Chloro-4-pyridyl)-3-(phenyl-d5)urea
IUPAC Name 1-(2-chloropyridin-4-yl)-3-(2,3,4,5,6-pentadeuteriophenyl)urea
SMILES [2H]c1c([2H])c([2H])c(NC(=O)Nc2ccnc(Cl)c2)c([2H])c1[2H]

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C12H10ClN3O/c13-11-8-10(6-7-14-11)16-12(17)15-9-4-2-1-3-5-9/h1-8H,(H2,14,15,16,17)/i1D,2D,3D,4D,5D
Formula Weight 252.0826
Chemical Name or Material Forchlorfenuron D5 (phenyl D5)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.