Learn More
Liebherr™ Forced-Air Refrigerator with Alarm
Use these refrigerators for general laboratory storage of temperature sensitive samples. Liebherr™ Forced-Air Refrigerators feature plastic coated grid shelves and plastic interior containers.
Brand: Liebherr™ GKV 5790
Description
- Choice of white or stainless steel exterior
- SwingLine door design with interchangeable hinges
- Ergonomic slimline handle
- Automatic defrost
- Dynamic cooling system
- Electronic control system
- External digital temperature display