Learn More
Cole-Parmer Food, Milk, and Dairy Tubing
Product Code. 30053239 Shop All Cole-Parmer Products
missing translation for 'orderingAttributeHoverText'
Diameter (English) Inner:
1 in.
1-1/2 in.
1-1/4 in.
1-3/4 in.
1/16 in.
1/2 in.
1/4 in.
1/8 in.
2 in.
3/16 in.
3/32 in.
3/4 in.
3/8 in.
5/16 in.
5/8 in.
7/16 in.
7/8 in.
Diameter (English) Outer:
1 in.
1-1/2 in.
1-1/4 in.
1-1/8 in.
1-3/8 in.
1/2 in.
1/4 in.
1/8 in.
11/16 in.
13/16 in.
15/16 in.
2 in.
2-1/2 in.
2-1/4 in.
3/16 in.
3/4 in.
3/8 in.
5/16 in.
5/32 in.
5/8 in.
7/16 in.
7/8 in.
9/16 in.
missing translation for 'unitSize'
15m
Product Code. 30053239
missing translation for 'mfr': Cole-Parmer 5012159
Only null left
Add to basket
Add to basket
Smooth, nonporous bore inhibits particle entrapment
Crystal clear for visual inspection and flow monitoring. Non-toxic, odor-free, and taste-free. Smooth, non-porous and flexible tubing promotes a sanitary fluid path. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Diameter (English) Inner 1/4 in.
Diameter (English) Outer 1/2 in.
Sterilization Method No
Color Clear