missing translation for 'onlineSavingsMsg'
Learn More

Fondenafil, TRC

Product Code. 30392672
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392672 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392672 Supplier Toronto Research Chemicals Supplier No. TRCF68540025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbodenafil
CAS 944241-52-5
Purity Grade Notes HPLC
Molecular Formula C24 H32 N6 O3
Quantity 25 mg
Synonym 7H-Pyrazolo[4,3-d]pyrimidin-7-one, 5-[2-ethoxy-5-[(4-ethyl-1-piperazinyl)carbonyl]phenyl]-1,6-dihydro-1-methyl-3-propyl-, Carbodenafil, Fondenafil
InChI Formula InChI=1S/C24H32N6O3/c1-5-8-18-20-21(28(4)27-18)23(31)26-22(25-20)17-15-16(9-10-19(17)33-7-3)24(32)30-13-11-29(6-2)12-14-30/h9-10,15H,5-8,11-14H2,1-4H3,(H,25,26,31)
SMILES CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)C(=O)N4CCN(CC)CC4
IUPAC Name 5-[2-ethoxy-5-(4-ethylpiperazine-1-carbonyl)phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molecular Weight (g/mol) 452.55
Formula Weight 452.2536
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.