missing translation for 'onlineSavingsMsg'
Learn More

Fondaparinux sodium, MedChemExpress

Product Code. 16481018
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481018 5 mg 5mg
16448548 10 mg 10mg
16423796 25 mg 25mg
16404417 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481018 Supplier Medchem Express Supplier No. HYB0597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor.

Fondaparinux sodium is an antithrombin-dependent factor Xa inhibitor.

Specifications

Chemical Name or Material Fondaparinux sodium
Physical Form Solid
CAS 114870-03-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C31H43N3Na10O49S8
For Use With (Application) COVID-19-immunoregulation
Synonym Fondaparin sodium SR-90107A
Solubility Information H2O : 125 mg/mL (72.33 mM; Need ultrasonic)
SMILES O[C@H]([C@@H](O)[C@@H]1O[C@](O[C@H](COS(=O)(O[Na])=O)[C@@H](O)[C@@H]2O)([H])[C@@H]2NS(=O)(O[Na])=O)[C@](O[C@@H]1C(O[Na])=O)([H])O[C@H]([C@H](O[C@@H]3O[C@]([C@@H]4O)([H])[C@@H](O[C@@H](O[C@]([C@@H]5O)([H])[C@H](O[C@H](OC)[C@@H]5NS(=O)(O[Na])=O)COS(=O)(O[Na])=O)[C@@H]4OS(=O)(O[Na])=O)C(O[Na])=O)COS(=O)(O[Na])=O)[C@@H]([C@H]3NS(=O)(O[Na])=O)OS(=O)(O[Na])=O
Molecular Weight (g/mol) 1728.08
Formula Weight 1728.08
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.