missing translation for 'onlineSavingsMsg'
Learn More

Fomblin YR-1800, Thermo Scientific Chemicals

Product Code. 11338496
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11338496 25 g 25g
11328496 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11338496 Supplier Thermo Scientific Alfa Aesar Supplier No. L17548.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

It is used in applications include pump and compressor cleaning, degreasing, dewatering, vapor blanketing, flash point reduction, and trace residue elimination. It acts as a diluent in the application of more viscous PFPE fluids and greases.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69991-67-9
Molecular Formula C3F6
Molecular Weight (g/mol) 150.023
MDL Number MFCD01323324
InChI Key HCDGVLDPFQMKDK-UHFFFAOYSA-N
Synonym hexafluoropropylene, hexafluoropropene, perfluoropropene, perfluoropropylene, propene, hexafluoro, perfluoro-1-propene, propylene, hexafluoro, hexfluoropropylene, 1-propene, 1,1,2,3,3,3-hexafluoro, 1,1,2,3,3,3-hexafluoro-1-propene
PubChem CID 8302
IUPAC Name 1,1,2,3,3,3-hexafluoroprop-1-ene
SMILES C(=C(F)F)(C(F)(F)F)F

Specifications

Density 1.92
Solubility Information Difficult to mix in water.
Quantity 25 g
Refractive Index 1.304
Chemical Name or Material Fomblin YR-1800

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.