missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOLR3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA537164
This item is not returnable.
View return policy
Description
This antibody detects endogenous protein at a molecular weight of 28 kDa. Purity is >95% by SDS-PAGE.
Folate is an essential vitamin that must be obtained from food intake through intestinal absorption in mammals. Folate and reduced folic acid derivatives bind to the folate receptor (FR) family, which mediates the endocytosis of 5-methyltetrahydrofolate into the cell. The folate receptors consist of five members, alpha, beta, gamma, gamma' (which is produced by alternative splicing) and delta. beta-FR is attached to the membrane by a GPI anchor and is expressed in malignant tissues of epithelial and nonepithelial origin. gamma-FR is expressed in tissues of hematopoietic origin, such as spleen, thymus and bone marrow. The expression pattern of delta-FR is elusive, which suggests that it is highly restricted both spatially and temporally. The alpha isoform of the folate receptor is used as a highly selective tumor marker and may be targeted for the delivery of therapeutic compounds to tumor cells by coupling to derivatives of folic acid.
Specifications
| FOLR3 | |
| Polyclonal | |
| Unconjugated | |
| FOLR3 | |
| Folate receptor 3; folate receptor 3 (gamma); folate receptor gamma; FOLR3; FR gamma; FR- gamma; FR-G; FR-gamma; gamma-hFR | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2352 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P41439 | |
| FOLR3 | |
| A synthetic peptide corresponding to residues in human FOLR3. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction