missing translation for 'onlineSavingsMsg'
Learn More

FOLR3 Antibody [Allophycocyanin], Novus Biologicals™

Product Code. 30735843 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30735843 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30735843 Supplier Novus Biologicals Supplier No. AF5319A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

FOLR3 Polyclonal antibody specifically detects FOLR3 in Human samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOLR3
Applications Western Blot, Flow Cytometry (Intracellular Staining), CyTOF
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias Folate receptor 3, folate receptor 3 (gamma), folate receptor gamma, FR-G, FR-gamma, gamma-hFR
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human FOLR3 long isoform, Ala20-Ser243, Accession # P41439
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2352
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.