missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR2 Monoclonal Antibody (EM-35)

FOLR2 Monoclonal Antibody (EM-35)
Brand: Invitrogen MA528552
This item is not returnable.
View return policy
Description
FOLR2 Monoclonal Antibody for Western Blot, ICC/IF, Flow, IP
Specifications
| FOLR2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15 mM sodium azide; pH 7.4 | |
| P14207 | |
| FOLR2 | |
| BW5147alpha,beta- cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| EM-35 | |
| Unconjugated | |
| FOLR2 | |
| BETA-HFR; FBP; FBP/PL-1; Fbp2; folate binding protein; folate binding protein 2; folate receptor 2; folate receptor 2 (fetal); folate receptor beta; folate receptor, beta; folate receptor, fetal/placental; Folate-binding protein 2; folate-binding protein, fetal/placental; Folbp2; Folbp-2; FOLR2; FR-beta; FR-P3; placental folate-binding protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 2350 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction