missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR1 Mouse anti-Human, Clone:2G5C12, Abnova™
Mouse monoclonal antibody raised against recombinant human FOLR1.
Brand: Abnova MAB17630.100ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the folate receptor (FOLR) family. Members of this gene family have a high affinity for folic acid and for several reduced folic acid derivatives, and mediate delivery of 5-methyltetrahydrofolate to the interior of cells. This gene is composed of 7 exons; exons 1 through 4 encode the 5′ UTR and exons 4 through 7 encode the open reading frame. Due to the presence of 2 promoters, multiple transcription start sites, and alternative splicing of exons, several transcript variants are derived from this gene. These variants differ in the lengths of 5′ and 3′ UTR, but they encode an identical amino acid sequence. [provided by RefSeq]Specifications
| FOLR1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunocytochemistry (1:100-1:500) Flow Cytometry (1:200-1:400) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. |
|
| FOLR1 | |
| Mouse | |
| 100 μg | |
| Human | |
| Antibody | |
| IgG2a |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 2G5C12 | |
| folate receptor 1 (adult) | |
| In PBS (0.05% sodium azide) | |
| FBP, FOLR, FR-alpha, MOv18 | |
| Recombinant protein corresponding to amino acid 41-227 of human FOLR1 from E. coli. | |
| 2348 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction