missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOLR1 Antibody (SF55 (BGN/1210/55)), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62577AF488
This item is not returnable.
View return policy
Description
FOLR1 Monoclonal specifically detects FOLR1 in Bovine samples. It is validated for ELISA.
Specifications
| FOLR1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FOLR1 | |
| Folate binding protein from bovine milk | |
| 0.1 mL | |
| Cancer | |
| 2348 | |
| Bovine | |
| Purified |
| ELISA | |
| SF55 (BGN/1210/55) | |
| ELISA | |
| Adult folate-binding protein, FBP, folate binding protein, Folate receptor 1, folate receptor 1 (adult), folate receptor alpha, Folate receptor, adult, FOLR, FR-alpha, KB cells FBP, Ovarian tumor-associated antigen MOv18 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| NB100-62577 reacts with folate binding protein (FBP), a 30 kDa protein with a high affinity for folate and several reduced folic acid derivatives that mediates the intracelullar delivery of 5 methyltetrahydrofolate. Membrane bound and soluble forms of a high affinity FBP have been found in kidney, placenta, serum, milk, and several cell lines. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction