missing translation for 'onlineSavingsMsg'
Learn More

Folpet, TRC

Product Code. 17961195
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17961195 25 g 25g
17924415 10 g 10g
17952375 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17961195 Supplier Toronto Research Chemicals Supplier No. TRCF40200025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-07-3
Molecular Formula C9 H4 Cl3 N O2 S
Molecular Weight (g/mol) 296.56
Synonym 2-[(Trichloromethyl)thio]-1H-isoindole-1,3(2H)-dione, Acryptane, Cosan T, Dipet, Faltan, Folnit, Folpan, Folpel, Ftalan, Fungitrol 11, Intercide TMP, N-[(Trichloromethyl)thio]phthalimide, Orthofaltan 50, Orthophaltan, Phaltan, Phaltane, Phthaltan, Ryl, Sanfol, Spolacid, Trifol, Vinicoli
IUPAC Name 2-(trichloromethylsulfanyl)isoindole-1,3-dione
SMILES ClC(Cl)(Cl)SN1C(=O)c2ccccc2C1=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H4Cl3NO2S/c10-9(11,12)16-13-7(14)5-3-1-2-4-6(5)8(13)15/h1-4H
Formula Weight 294.9028
Percent Purity >95%
Chemical Name or Material Folpet
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.