missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Folliculin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29210
This item is not returnable.
View return policy
Description
Folliculin Polyclonal antibody specifically detects Folliculin in Human samples. It is validated for Immunohistochemistry
Specifications
| Folliculin | |
| Polyclonal | |
| Tris-buffered Saline 0.1% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry | |
| Unconjugated | |
| BHD skin lesion fibrofolliculoma protein, BHDFLCL, Birt-Hogg-Dube syndrome protein, DKFZp547A118, FLJ45004, FLJ99377, folliculin, MGC17998, MGC23445 | |
| Between 1 and 50 | |
| 25 μg | |
| Cancer, Cell Cycle and Replication | |
| 201163 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction