missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Follicle Stimulating Hormone Recombinant Rabbit Monoclonal Antibody (6R1V3)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558229
This item is not returnable.
View return policy
Description
Immunogen sequence: APDVQDCPEC TLQENPFFSQ PGAPILQCMG CCFSRAYPTP LRSKKTMLVQ KNVTSESTCC VAKSYNRVTV MGGFKVENHT ACHCSTCYYH KS.
Follicle-stimulatine hormone (FSH) is a member of the glycoprotein hormone family. Members of this family consist of an identical alpha subunit and a hormone-specific beta subunit. Alternative splicing results in two transcript variants encoding the same protein. FSH is secreted by gonadotropic cells in the pituitary gland and in conjunction with luteinizing hormone, induces egg and sperm production.
Specifications
| Follicle Stimulating Hormone | |
| Recombinant Monoclonal | |
| 0.79 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P01225, Q60687 | |
| FSHB | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 25-116 of human FSH alpha/beta (NP_000726.1/NP_000501.1). | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| 6R1V3 | |
| Unconjugated | |
| FSHB | |
| class 1 follicle-stimulating hormone beta polypeptide; class 2 follicle-stimulating hormone beta polypeptide; follicle - stimulating hormone subunit beta; follicle stimulating hormone beta; follicle stimulating hormone beta subunit; follicle stimulating hormone beta-subunit precursor; follicle stimulating hormone subunit beta; follicle stimulating hormone, beta polypeptide; follicle-stimulating hormone beta subunit; follicle-stimulating hormone beta-subunit; follicle-stimulating hormone subunit beta; follicule stimulating hormone, beta polypeptide; follitropin beta chain; Follitropin subunit beta; follitropin, beta chain; FSH; FSH-alpha; FSHB; FSH-B; Fshbeta; FSH-beta; HH24; LSH-alpha; prefollicle stimulating hormone beta chain; TSH-alpha | |
| Rabbit | |
| Protein A | |
| RUO | |
| 14308, 2488 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction