missing translation for 'onlineSavingsMsg'
Learn More

Folinic Acid Calcium Salt, TRC

Product Code. 30135024
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135024 250 mg 250mg
30135545 500 mg 500mg
17951065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135024 Supplier Toronto Research Chemicals Supplier No. TRCF680320250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20 H21 N7 O7 . Ca
Molecular Weight (g/mol) 511.5
Synonym Calcium folinate, Calcium (2S)-2-[[4-[[[(6RS)-2-amino-5-formyl-4-oxo1,4,5,6,7,8-hexahydropteridin-6-yl]methyl]amino]benzoyl]amino]pentanedioate, Calcium 5-formyltetrahydrofolate, Calcium L-folinate, Calcium folinate, Calcium leucovorin, Folidan, Folinic acid calcium salt, Lederfoline, Leucovorin calcium, Leucovorin calcium salt, NSC 3590, Rescuvolin, Wellcovorin
IUPAC Name calcium;(2S)-2-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]pentanedioate
SMILES [Ca+2].NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-])N2C=O)N1

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1
Formula Weight 511.1128
Percent Purity >95%
Chemical Name or Material Calcium Folinate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.