missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid calcium salt pentahydrate, MedChemExpress

Product Code. 16422006
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16422006

Brand: Medchem Express HYB0080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Folinic acid calcium salt pentahydrate (Leucovorin calcium salt pentahydrate) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.

Folinic acid calcium salt pentahydrate (Leucovorin calcium salt pentahydrate) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.

Specifications

Chemical Name or Material Folinic acid calcium salt pentahydrate
Physical Form Solid
CAS 6035-45-6
Color Light Yellow
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C20H31CaN7O12
For Use With (Application) Cancer-programmed cell death
Synonym Leucovorin calcium salt pentahydrate
Solubility Information H2O : 10 mg/mL (16.62 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble)
SMILES O=CN1C(C2=O)=C(NC(N)=N2)NCC1CNC3=CC=C(C(N[C@H](C([O-])=O)CCC([O-])=O)=O)C=C3.O.O.O.O.[Ca+2].O
Molecular Weight (g/mol) 601.58
Formula Weight 601.58
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.