missing translation for 'onlineSavingsMsg'
Learn More

Folinic acid, calcium salt, 99.4%, MP Biomedicals™

Product Code. 11464742 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 mg
0.1 g
0.5 g
1 g
Unit Size:
100mg
1g
25mg
500mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464742 0.5 g 500mg
11444742 25 mg 25mg
11454742 0.1 g 100mg
11474742 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11464742 Supplier MP Biomedicals Supplier No. 0219150490
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 1492-18-8
Molecular Formula C20H21CaN7O7
Molecular Weight (g/mol) 511.51
MDL Number MFCD00006704
InChI Key KVUAALJSMIVURS-AEQORCIRNA-L
Synonym calcium folinate
PubChem CID 131675590
SMILES [Ca++].NC1=NC(=O)C2=C(NCC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC([O-])=O)C([O-])=O)N2C=O)N1

Specifications

Color Yellow
Assay Percent Range 0.994
Quantity 0.5 g
IUPAC Name calcium (2S)-2-[(4-{[(2-amino-5-formyl-4-oxo-1,4,5,6,7,8-hexahydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioate
Formula Weight 511.5
Physical Form Powder
Chemical Name or Material Folinic acid, calcium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.