missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folinic acid calcium, MedChemExpress
Description
Folinic acid calcium (Leucovorin calcium) is a biological folic acid and is generally administered along with methotrexate (MTX) as a rescue agent to decrease MTX-induced toxicity.
Specifications
Specifications
| Chemical Name or Material | Folinic acid calcium |
| Physical Form | Solid |
| CAS | 1492-18-8 |
| Color | Light Yellow |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C20H21CaN7O7 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Leucovorin calcium Calcium folinate |
| Solubility Information | H2O : 50 mg/mL (97.75 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?