missing translation for 'onlineSavingsMsg'
Learn More

Folinate calcium 100 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30318277
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30318277 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318277 Supplier Dr. Ehrenstorfer Supplier No. DREA13888500WA100
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Calcium Folinate
CAS 1492-18-8
Analyte or Component Names Calcium Folinate
Shipping Condition Room Temperature
Molecular Formula C20 H21 N7 O7 . Ca
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Calcium folinate, Calcium (2S)-2-[[4-[[[(6RS)-2-amino-5-formyl-4-oxo1,4,5,6,7,8-hexahydropteridin-6-yl]methyl]amino]benzoyl]amino]pentanedioate, Calcium 5-formyltetrahydrofolate, Calcium L-folinate, Calcium folinate, Calcium leucovorin, Folidan, Folinic acid calcium salt, Lederfoline, Leucovorin calcium, Leucovorin calcium salt, NSC 3590, Rescuvolin, Wellcovorin, Folic Acid Related Compound A as Calcium Salt
InChI Formula InChI=1S/C20H23N7O7.Ca/c21-20-25-16-15(18(32)26-20)27(9-28)12(8-23-16)7-22-11-3-1-10(2-4-11)17(31)24-13(19(33)34)5-6-14(29)30;/h1-4,9,12-13,22H,5-8H2,(H,24,31)(H,29,30)(H,33,34)(H4,21,23,25,26,32);/q;+2/p-2/t12?,13-;/m0./s1
SMILES [Ca+2].NC1=NC(=O)C2=C(NCC(CNc3ccc(cc3)C(=O)N[C@@H](CCC(=O)[O-])C(=O)[O-])N2C=O)N1
IUPAC Name calcium;(2S)-2-[[4-[(2-amino-5-formyl-4-oxo-1,6,7,8-tetrahydropteridin-6-yl)methylamino]benzoyl]amino]pentanedioate
Solvent or Matrix Water
Molecular Weight (g/mol) 511.5
Quantity 1 mL
Formula Weight 511.1128 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.