missing translation for 'onlineSavingsMsg'
Learn More

Folin & Ciocalteu'S pHenol Reagent, Pure, Fisher Chemical™

Product Code. 10191520 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10191520 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10191520 Supplier Fisher Chemical Supplier No. J/4100/08
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Folin & Ciocalteu'S pHenol Reagent, Pure, Fisher Chemical, Quantity: 500mL, Packaging: Amber glass bottle, Yellow, MDL Number: 132625, Physical Form: Liquid

Folin & Ciocalteu'S pHenol Reagent, Pure, Fisher Chemical, Quantity: 500mL, Packaging: Amber glass bottle, Yellow, MDL Number: 132625, Physical Form: Liquid

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
MDL Number 132625
Packaging Amber glass bottle
Quantity 500 mL
Physical Form Liquid
compliance-icons
Product Identifier
  • Folin & Ciocalteu's phenol reagent
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.