missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, MedChemExpress

Product Code. 16409647
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409647 5 g 5g
16457098 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16409647 Supplier Medchem Express Supplier No. HY16637

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Folic acid (Vitamin B9) is a orally active essential nutrient from the B complex group of vitamins. Folic acid shows antidepressant-like effect. Folic acid sodium reduces the risk of neonatal neural tube defects. Folic acid can be used to the treatment of megaloblastic and macrocytic anemias due to folic deficiency.

Folic acid (Vitamin B9) is a orally active essential nutrient from the B complex group of vitamins. Folic acid shows antidepressant-like effect. Folic acid sodium reduces the risk of neonatal neural tube defects. Folic acid can be used to the treatment of megaloblastic and macrocytic anemias due to folic deficiency.

Specifications

Chemical Name or Material Folic acid
CAS 59-30-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H19N7O6
Quantity 5 g
Synonym Vitamin B9 Vitamin M
Solubility Information 1M NaOH : 100 mg/mL (226.55 mM; Need ultrasonic) ∣DMSO : 33.33 mg/mL (75.51 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(O)CC[C@@H](C(O)=O)NC(C1=CC=C(NCC2=CN=C3N=C(N)NC(C3=N2)=O)C=C1)=O
Molecular Weight (g/mol) 441.4
Formula Weight 441.4
Percent Purity 98.01%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.