missing translation for 'onlineSavingsMsg'
Learn More

Folic Acid-d4, TRC

Product Code. 30400468
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400468 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30400468 Supplier Toronto Research Chemicals Supplier No. TRCF6803025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Folic Acid-d4
CAS 171777-72-3
Purity Grade Notes HPLC
Molecular Formula C19 2H4 H15 N7 O6
Quantity 5 mg
Synonym L-Glutamic acid, N-[4-[[(2-amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl-2,3,5,6-d4]-, L-Glutamic acid, N-[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl-2,3,5,6-d4]- (9CI), N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl]amino]benzoyl-2,3,5,6-d4]-L-glutamic acid, Folic acid-d4, Folic acid D4 (benzoyl-2,3,5,6-D4), Vitamin B9-d4
InChI Formula InChI=1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c(C(=O)N[C@@H](CCC(=O)O)C(=O)O)c([2H])c([2H])c1NCc2cnc3nc(N)nc(O)c3n2
IUPAC Name (2S)-2-[[4-[(2-amino-4-hydroxypteridin-6-yl)methylamino]-2,3,5,6-tetradeuteriobenzoyl]amino]pentanedioic acid
Molecular Weight (g/mol) 445.42
Formula Weight 445.1648
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.