missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folic Acid-d2, TRC
Specifications
Specifications
| Chemical Name or Material | Folic Acid-d2 |
| CAS | 69022-87-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19H17D2N7O6 |
| Quantity | 5 mg |
| Synonym | L-Glutamic acid, N-[4-[[(2-amino-1,4-dihydro-4-oxo-6-pteridinyl)methyl-d2]amino]benzoyl]- (9CI), [9,9-2H]Folic acid, N-[4-[[(2-Amino-3,4-dihydro-4-oxo-6-pteridinyl)methyl-d2]amino]benzoyl]-L-glutamic acid, Folic acid-d2, Folic acid D2 (aminomethylene-D2), Vitamin B9-d2 |
| InChI Formula | InChI=1S/C19H19N7O6/c20-19-25-15-14(17(30)26-19)23-11(8-22-15)7-21-10-3-1-9(2-4-10)16(29)24-12(18(31)32)5-6-13(27)28/h1-4,8,12,21H,5-7H2,(H,24,29)(H,27,28)(H,31,32)(H3,20,22,25,26,30)/t12-/m0/s1/i7D2 |
| SMILES | [2H]C([2H])(Nc1ccc(cc1)C(=O)N[C@@H](CCC(=O)O)C(=O)O)c2cnc3NC(=NC(=O)c3n2)N |
| Molecular Weight (g/mol) | 443.41 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?