missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, crystalline

Product Code. p-7041956
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15443539

Brand: Thermo Scientific Alfa Aesar J60833.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-30-3 | C19H19N7O6 | 441.404 g/mol

Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid acts as a vitamin (hematopoietic), a nutritional supplement and dietary requirement for poultry. It also plays an important role in the formation of S-adenosyl methionine and in the synthesis and repair of DNA and RNA. Further, it serves as an essential vitamin for erythropoiesis. It is also employed in the treatment of methanol poisoning. In addition to this, it is used in the treatment and prevention of folate deficiencies and megaloblastic anemia.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cellsFolic acid acts as a vitamin (hematopoietic), a nutritional supplement and dietary requirement for poultry. It also plays an important role in the formation of S-adenosyl methionine and in the synthesis and repair of DNA and RNA. Further, it serves as an essential vitamin for erythropoiesis. It is also employed in the treatment of methanol poisoning. In addition to this, it is used in the treatment and prevention of folate deficiencies and megaloblastic anemia.

Solubility
Soluble in sodium hydroxide, acetic acid, phenol, pyridine, alkali hydroxides, carbonates, hot dilute hydrochloric acid and sulfuric acid. Slightly soluble in water, methanol, ethanol and butanol. Insoluble in acetone, chloroform, ether and benzene.

Notes
Light and heat sensitive. Incompatible with heavy metal ions, strong oxidizing agents and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Color Yellow to Orange
Quantity 25 g
Beilstein 100781
Merck Index 14,4221
Solubility Information Soluble in sodium hydroxide,acetic acid,phenol,pyridine,alkali hydroxides,carbonates,hot dilute hydrochloric acid and sulfuric acid. Slightly soluble in water,methanol,ethanol and butanol. Insoluble in acetone,chloroform,ether and benzene.
Formula Weight 441.41
Physical Form Crystalline
Chemical Name or Material Folic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.