missing translation for 'onlineSavingsMsg'
Learn More

Folic acid, 95-102%

Product Code. 15455239
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15455239

Brand: Thermo Scientific Alfa Aesar J62937.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59-30-3 | C19H19N7O6 | 441.404 g/mol

Folic acid and its derivatives are essential mediators of one-carbon metabolism within cells.It is especially important in aiding rapid cell division and growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-30-3
Molecular Formula C19H19N7O6
Molecular Weight (g/mol) 441.404
MDL Number MFCD00079305
InChI Key OVBPIULPVIDEAO-LBPRGKRZSA-N
Synonym folic acid, folate, pteroylglutamic acid, vitamin m, folacin, folacid, pteglu, vitamin b9, folvite, pteroyl-l-glutamic acid
PubChem CID 6037
ChEBI CHEBI:27470
IUPAC Name (2S)-2-[[4-[(2-amino-4-oxo-1H-pteridin-6-yl)methylamino]benzoyl]amino]pentanedioic acid
SMILES C1=CC(=CC=C1C(=O)NC(CCC(=O)O)C(=O)O)NCC2=CN=C3C(=N2)C(=O)N=C(N3)N

Specifications

Packaging Glass bottle
Quantity 100 g
Beilstein 100781
Solubility Information Soluble in water (1.6mg/ml),1 M NaOH (50mg/ml),methanol (slightly),diluted acids,and alkaline solutions.
Formula Weight 441.41
Physical Form Powder
Chemical Name or Material Folic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.