missing translation for 'onlineSavingsMsg'
Learn More

Getinge™ Folded Tissues

Product Code. 13433906
Encompass
Click to view available options
Packaging Quantity:
20 Pk. x 160
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13433906

Brand: Getinge™ 1152751

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Contains 160 sheets per pack. Getinge™ Getinge™ Folded Tissues prevent unnecessary waste that often comes with rolled tisssue.

  • 320 x 220mm
  • Each pack contains 160 sheets

Specifications

Product Type Tissue
Length (Metric) 220 mm
Width (Metric) 320 mm
Format Folded sheet
Packaging Quantity 20 Pk. x 160
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.