Description
General Features
- Individually packed
- Color: white
- With exhalation valve
Safety Features
- Certified FFP3 face mask to EN 149
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
- Soft sealing lip in the nose area guarantees a secure, comfortable sealed position of the mask
- Adjustable head band offering easy handling and comfortable fit
- Flexible and adjustable nose clip
- Soft material edges for high wearing comfort
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 149 (D marked) |
| Disposable | Single-use |
| Material | Polypropylene, Polyester |
| With Exhalation Valve | Yes |
| Filter Efficiency | FFP3 |
| Sterility | Non-sterile |