Learn More
Uvex Fold-flat Mask silv-Air p 8313 FFP3
Product Code. 15630285 Shop All Uvex Products
Click to view available options
:
Filter Type: FFP3
Product Code. 15630285
Brand: Uvex 8708313
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The gray coloring on the outside of the silv-Air p masks prevents them from looking dirty quickly and having to be disposed of prematurely by the user. By contrast, the inside is of course pure white. In addition and with all products in this family, the 3D nose geometry ensures an optimum fit, even with different face shapes.
General Features
  • Particle-filtering flat-fold mask with climate-optimized exhalation valve
  • Hydrophobic material coating
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Flexible, adjustable integrated nose clip
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Individually sealed
  • Color: silver
Protection Features
  • FFP3 respirator certified in accordance with EN 149:2001+A1:2009
  • D marking (mask has passed the dolomite dust test): good breathability even with high levels of dust
  • Not reusable
Comfort Features
  • Four-point headband attachment for a comfortable, individual and secure fit
  • Flexible, adjustable integrated nose clip
  • Very soft, all-around sealing lip for optimal wearer comfort and secure fit
  • Exhalation valve to reduce breathing resistance and minimise heat and moisture build-up inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type R - Somewhat Resistant to Oil
Certifications/Compliance (Europe) EN 149 (D marked)
Disposable Single-use
Material Polypropylene, Polyester
Size Standard
With Exhalation Valve Yes
Filter Efficiency FFP3
Sterility Non-sterile