Learn More
Uvex Fold-flat Mask silv-Air c 3210 FFP2
Product Code. 17572965 Shop All Uvex Products
Change view
Click to view available options
:
Filter Type: FFP2
Product Code.
17572965 Filter Type: FFP2
1 options
Product Code. 17572965 Supplier Uvex Supplier No. 8733210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
The silv-Air c product family consists of classic folding and cup-shaped face masks which fit different face shapes. Headband lacing system and integrated sealing lips in all protection classes make it easy to fit these masks correctly – and to wear them comfortably for long periods.
General Features
  • Particle-filtering folding mask with exhalation valve
  • Color: white
Protection Features
  • Certified FFP2 face mask to EN 149:2001+A1:2009
  • D label (dolomite dust test passed): maximum breathability even with really high dust levels
  • Cannot be reused
Comfort Features
  • Seamless headband and flexible, adjustable nose clip
  • Comfortable sealing lip in nose area for secure fit
  • Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 149 (D marked)
Design Type Flat-fold
Disposable Single-use
Material Polypropylene, Polyester
Size Standard
With Exhalation Valve Yes
Filter Efficiency FFP2
Sterility Non-sterile