missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TLC 1598 Thermometer
GREENER_CHOICE

Product Code. 10727421
Click to view available options
Max. Temperature (Metric):
200°C
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10727421

Brand: Ebro Electronic™ 13401620A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Thermometer with fold-back probe

Temperature logger with external probe.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012; EN 13485; (EC) 1935/2004 Materials and articles which come into contact with foodstuffs; The Food and Feed Hygiene and Safety Regulations 2020 2020 No. 1504; PART 3; Regulation 16
Disposable Single-use
For Use With (Application) Precision Fold-Back Thermometer with foldable Pt 1000 penetration probe and high accuracy
Height (Metric) 38 mm
Includes Thermometer, calibration certificate, instruction manual
Length (Metric) 190 mm
Material Stainless Steel, Plastic
Model TLC 1598
Probe Type PT1000
Resolution 0.1°C
Weight (Metric) 0.085 kg
Data Storage None
Max. Temperature (English) 392°F
Max. Temperature (Metric) 200°C
Min. Temperature (English) -58°F
Min. Temperature (Metric) -50°C
Calibrated Factory Calibrated
For Use With (Equipment) AG 121
Battery Life Approx. 4 years
Battery Type Lithium 3 V/Typ CR 2450
Accuracy ±0.3@deg;C
Immersion Length 120 mm
Display LCD
Width (Metric) 120 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.