missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Folcisteine, MedChemExpress
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Description
Folcisteine is a plant growth regulator.
Specifications
Specifications
| Chemical Name or Material | Folcisteine |
| Physical Form | Solid |
| CAS | 5025-82-1 |
| Color | White |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C6H9NO3S |
| Synonym | 3-Acetylthiazolidine-4-carboxylic acid |
| Solubility Information | DMSO : ≥ 160 mg/mL (913.19 mM) |
| SMILES | O=C(C1N(C(C)=O)CSC1)O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction