missing translation for 'onlineSavingsMsg'
Learn More

Folate Receptor alpha Monoclonal Antibody (548908), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA523917

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15503612

  • £378.00 / 100µg
Estimated Shipment: 15-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

In direct ELISAs, no cross-reactivity with recombinant human FOLR2, 3 or 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Folate Receptor alpha Monoclonal antibody specifically detects Folate Receptor alpha in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Western Blot.
Specifications

Specifications

Folate Receptor alpha
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P15328
FOLR1
CHO-derived recombinant human FOLR1 Arg25-Met233
100 μg
Primary
Human
Antibody
IgG1
ELISA, Flow Cytometry, Immunocytochemistry, Western Blot
548908
Unconjugated
FOLR1
66/67kDa; AA407891; adult folate-binding protein; eIF3 p66; EIF3D; eIF3d {ECO:0000255; eIF3p66; eIF3-p66; EIF3S7; eIF-3-zeta; eIF3-zeta; eIF-3-zeta {ECO:0000255; Eukaryotic translation initiation factor 3 subunit 7; eukaryotic translation initiation factor 3 subunit 7 {ECO:0000255; eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3 subunit D {ECO:0000255; eukaryotic translation initiation factor 3, subunit 7 (zeta); eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; FBP; Fbp1; folate binding protein; folate binding protein 1; folate receptor 1; folate receptor 1 (adult); folate receptor alpha; Folate receptor, adult; folate-binding protein 1; Folbp1; Folbp-1; FOLR; FOLR1; FR-alpha; HAMAP-Rule:MF_03003}; KB cells FBP; MOv18; Ovarian tumor-associated antigen MOv18; translation initiation factor eIF3 p66; translation initiation factor eIF3 p66 subunit
Mouse
Protein A/G
RUO
2348
-20° C, Avoid Freeze/Thaw Cycles
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers