missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOG2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32895
This item is not returnable.
View return policy
Description
FOG2 Polyclonal specifically detects FOG2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| FOG2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| transcription factor GATA4, modulator of, zinc finger protein 89B, zinc finger protein, multitype 2 | |
| Rabbit | |
| 128 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| 0.96mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8WW38 | |
| ZFPM2 | |
| Recombinant protein encompassing a sequence within the center region of human FOG2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 23414 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction