Learn More
Leica Microsystems Focus Column for S4/S6/S8 StereoZoom Microscope
Description
- Designed for S4/S6/S8 stereo zoom microscopes.
- Replacement kit includes Allen key and three hex-head screws.
Specifications
Specifications
| For Use With (Equipment) | S4/S6/S8 microscopes |
| Includes | Allen key and three hex-head screws. |