missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNTB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106707
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total FNTB.
Eukaryotic cells contain 3 different types of prenyltransferases that attach either a farnesyl group (15 carbons) or a geranylgeranyl group (20 carbons) in thioether linkage to C-terminal cysteine residues in a variety of proteins. These posttranslational modifications provide a mechanism for membrane localization of proteins that lack a transmembrane domain. CAAX farnesyltransferase (FTase) attaches a farnesyl group from farnesyl pyrophosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box, where C is cysteine, A is usually but not always an aliphatic amino acid, and X is typically methionine or serine. This enzyme has the ability to farnesylate peptides as short as 4 residues in length that conform to the CAAX consensus sequence. The gene for the beta subunit of CAAX farnesyltransferase (FNTB) has been pinpointed to 14q23-q24 by Southern blot hybridization and PCR analyses of panels of human/Chinese hamster somatic cell hybrid lines and by fluorescence chromosomal in situ hybridization.
Specifications
| FNTB | |
| Polyclonal | |
| Unconjugated | |
| FNTB | |
| 2010013E13Rik; AA409500; CAAX farnesyltransferase subunit beta; EC 2.5.1.58; farnesyl-protein transferase beta-subunit; farnesyltransferase beta subunit; farnesyltransferase, CAAX box, beta; FNTB; FPTB; ft beta; FTase beta; FTase-beta; LOW QUALITY PROTEIN: protein farnesyltransferase subunit beta; protein farnesyltransferase subunit beta; Ras proteins prenyltransferase subunit beta | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 110606, 2342, 64511 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P49356, Q02293, Q8K2I1 | |
| FNTB | |
| A synthesized peptide derived from human FNTB(Accession P49356), corresponding to amino acid residues C254-Q304. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction