missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNTB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597757
This item is not returnable.
View return policy
Description
FNTB Polyclonal Antibody for ELISA, ICC/IF, IHC (P), IP, Western Blot
Eukaryotic cells contain 3 different types of prenyltransferases that attach either a farnesyl group (15 carbons) or a geranylgeranyl group (20 carbons) in thioether linkage to C-terminal cysteine residues in a variety of proteins. These posttranslational modifications provide a mechanism for membrane localization of proteins that lack a transmembrane domain. CAAX farnesyltransferase (FTase) attaches a farnesyl group from farnesyl pyrophosphate to cysteine residues at the fourth position from the C terminus of proteins that end in the CAAX box, where C is cysteine, A is usually but not always an aliphatic amino acid, and X is typically methionine or serine. This enzyme has the ability to farnesylate peptides as short as 4 residues in length that conform to the CAAX consensus sequence. The gene for the beta subunit of CAAX farnesyltransferase (FNTB) has been pinpointed to 14q23-q24 by Southern blot hybridization and PCR analyses of panels of human/Chinese hamster somatic cell hybrid lines and by fluorescence chromosomal in situ hybridization.
Specifications
| FNTB | |
| Polyclonal | |
| Unconjugated | |
| FNTB | |
| 2010013E13Rik; AA409500; CAAX farnesyltransferase subunit beta; EC 2.5.1.58; farnesyl-protein transferase beta-subunit; farnesyltransferase beta subunit; farnesyltransferase, CAAX box, beta; FNTB; FPTB; ft beta; FTase beta; FTase-beta; LOW QUALITY PROTEIN: protein farnesyltransferase subunit beta; protein farnesyltransferase subunit beta; Ras proteins prenyltransferase subunit beta | |
| Rabbit | |
| Protein G | |
| RUO | |
| 2342 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
| 3 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P49356 | |
| FNTB | |
| Recombinant Human Protein farnesyltransferase subunit beta protein (1-300AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction