missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNTB Mouse anti-Human, Mouse, Clone: 1F3B10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66783-1-IG-150UL
This item is not returnable.
View return policy
Description
ZNF24 is a new candidate transcription factor.Specifications
| FNTB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49356, Q8K2I1 | |
| FNTB | |
| FNTB Fusion Protein Ag27796 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| Western Blot | |
| 1F3B10 | |
| Unconjugated | |
| FNTB | |
| FNTB, FPTB, FTase beta | |
| Mouse | |
| Protein G | |
| RUO | |
| 110606, 2342 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction