missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ FNIP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29189
This item is not returnable.
View return policy
Description
FNIP1 Polyclonal antibody specifically detects FNIP1 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| FNIP1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| DKFZp781P0215, folliculin interacting protein 1, folliculin-interacting protein 1, KIAA1961DKFZp686E18167, MGC667 | |
| between 775 and 825 | |
| 100 μg | |
| Cell Biology | |
| 96459 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction