missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNDC5 Recombinant Rabbit Monoclonal Antibody (K01_1R23)
Description
FNDC5 Recombinant Monoclonal Antibody for Western Blot
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
| Antigen | FNDC5 |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | K01_1R23 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM tris glycine with 150mM NaCl, 40% glycerol, 0.05% BSA and 0.01% sodium azide; pH 7.4 |
| Gene | FNDC5 |
| Gene Accession No. | Q8K3V5, Q8K4Z2, Q8NAU1 |
| Gene Alias | 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?