missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FNDC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA581569
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse Irisin/FNDC5.
Fibronectin type III domain-containing protein 5, the precursor of irisin, is a protein that is encoded by the FNDC5 gene. This gene encodes a secreted protein that is released from muscle cells during exercise. The encoded protein may participate in the development of brown fat. Translation of the precursor protein initiates at a non-AUG start codon at a position that is conserved as an AUG start codon in other organisms. Alternative splicing results in multiple transcript variants.
Specifications
| FNDC5 | |
| Polyclonal | |
| Unconjugated | |
| FNDC5 | |
| 1500001L03Rik; AI836596; C87088; fibronectin type III domain containing 5; fibronectin type III domain-containing protein 5; Fibronectin type III repeat-containing protein 2; Fndc5; Frcp2; Irisin; PeP; Peroxisomal protein; Pxp | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 384061 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8K4Z2 | |
| FNDC5 | |
| A synthetic peptide corresponding to the center region of the Mouse FNDC5. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction