missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FNDC3A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-88809
This item is not returnable.
View return policy
Description
FNDC3A Polyclonal antibody specifically detects FNDC3A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
| FNDC3A | |
| Polyclonal | |
| Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| bA203I16.1, bA203I16.5, fibronectin type III domain containing 3, fibronectin type III domain containing 3A, fibronectin type-III domain-containing protein 3A, FNDC3, HUGO, Human gene expressed in odontoblasts, KIAA0970FLJ31509 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 22862 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| FNDC3A | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GAGDMTTQYMPQYQSSQVYGDVDAHSTHGRSNFRDERSSKTYERLQKKLKDRQGTQKDKMSSPPSSPQKCPSPINEHNGLIKGQIAGGINTGSAKIKSGKGKGGTQVDTEIEEKDEETKAFEALLSNIVKPVA | |
| 0.1 mL | |
| Primary | |
| Specificity of human FNDC3A antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction